Adrenocorticotropic Hormone Fragment 18-39 human
Synonym(s):
CLIP
Empirical Formula (Hill Notation):
C112H165N27O36
CAS Number:
Molecular Weight:
2465.67
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing