Skip to Content
Merck
Adenosine 5′-[γ-thio]triphosphate tetralithium salt ≥75% (HPLC), powder

Adenosine 5′-[γ-thio]triphosphate tetralithium salt

Adenosine 5′-[γ-thio]triphosphate tetralithium salt ≥75% (HPLC), powder
Synonym(s):
ATP-γ-S, Adenosine 5′-(3-thiotriphosphate) tetralithium salt, Ado-5′-PPP[S]
Empirical Formula (Hill Notation):
C10H12Li4N5O12P3S
CAS Number:
Molecular Weight:
546.98
EC Number:

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing