![Acetaminophen meets USP testing specifications, 98.0-102.0%, powder](/deepweb/assets/sigmaaldrich/product/structures/321/572/5924e2dc-ae59-47a1-b7ae-ff9e7c9254ac/640/5924e2dc-ae59-47a1-b7ae-ff9e7c9254ac.png)
Acetaminophen
![Acetaminophen meets USP testing specifications, 98.0-102.0%, powder](/deepweb/assets/sigmaaldrich/product/structures/321/572/5924e2dc-ae59-47a1-b7ae-ff9e7c9254ac/640/5924e2dc-ae59-47a1-b7ae-ff9e7c9254ac.png)
Synonym(s):
Acetaminophen, 4′-Hydroxyacetanilide, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, APAP, Paracetamol
Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Products
Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing