Skip to Content
Merck
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F2α solution, 10 mg/mL in methyl acetate

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F2α solution, 10 mg/mL in methyl acetate
Synonym(s):
U-46619, U46619
Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1