Skip to Content
Merck
7-Ethoxycoumarin analytical standard

7-Ethoxycoumarin

7-Ethoxycoumarin analytical standard
Synonym(s):
7-Ethoxy-1-benzopyran-2-one
Empirical Formula (Hill Notation):
C11H10O3
CAS Number:
Molecular Weight:
190.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing