Skip to Content
Merck
6-Maleimidohexanoic acid N-hydroxysuccinimide ester ≥98.0% (HPLC)

6-Maleimidohexanoic acid N-hydroxysuccinimide ester

6-Maleimidohexanoic acid N-hydroxysuccinimide ester ≥98.0% (HPLC)
Synonym(s):
6-Maleimidocaproic acid N-succinimidyl ester, N-(ε-Maleimidocaproyloxy)succinimide, N-Succinimidyl 6-maleimidocaproate, EMCS
Empirical Formula (Hill Notation):
C14H16N2O6
CAS Number:
Molecular Weight:
308.29

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing