Skip to Content
Merck
6-Amino-3-pyridinecarbonitrile 97%

6-Amino-3-pyridinecarbonitrile

6-Amino-3-pyridinecarbonitrile 97%
Synonym(s):
2-Amino-5-cyanopyridine, 6-Aminonicotinonitrile
Linear Formula:
C5H3NNH2CN
CAS Number:
Molecular Weight:
119.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing