
5′-Iodo-5′-deoxyadenosine

Empirical Formula (Hill Notation):
C10H12IN5O3
CAS Number:
Molecular Weight:
377.14
Products
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing