Skip to Content
Merck
4-Nitroaniline analytical standard

4-Nitroaniline

4-Nitroaniline analytical standard
Synonym(s):
4-Nitroaniline
Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing