Skip to Content
Merck
4-Androstenedione ≥98% (HPLC)

4-Androstenedione

4-Androstenedione ≥98% (HPLC)
Synonym(s):
4-Androstene-3,17-dione, Androstenedione
Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing