Skip to Content
Merck
4,5-Dichloroimidazole ≥98%

4,5-Dichloroimidazole

4,5-Dichloroimidazole ≥98%
Empirical Formula (Hill Notation):
C3H2Cl2N2
CAS Number:
Molecular Weight:
136.97

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing