Skip to Content
Merck
4,4-Pentamethylene-2-pyrrolidinone 96%

4,4-Pentamethylene-2-pyrrolidinone

4,4-Pentamethylene-2-pyrrolidinone 96%
Synonym(s):
3-Azaspiro[4.5]decan-2-one
Empirical Formula (Hill Notation):
C9H15NO
CAS Number:
Molecular Weight:
153.22

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing