![3-Phenyl-2-propyn-1-amine hydrochloride 97%](/deepweb/assets/sigmaaldrich/product/structures/182/819/3822aec7-69fb-4f08-a6f1-ceb91ad42e0c/640/3822aec7-69fb-4f08-a6f1-ceb91ad42e0c.png)
3-Phenyl-2-propyn-1-amine hydrochloride
![3-Phenyl-2-propyn-1-amine hydrochloride 97%](/deepweb/assets/sigmaaldrich/product/structures/182/819/3822aec7-69fb-4f08-a6f1-ceb91ad42e0c/640/3822aec7-69fb-4f08-a6f1-ceb91ad42e0c.png)
Synonym(s):
3-Phenylpropargylamine hydrochloride
Empirical Formula (Hill Notation):
C9H10ClN
CAS Number:
Molecular Weight:
167.64
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing