Skip to Content
Merck
3-Methoxypropionitrile ≥98.0% (GC)

3-Methoxypropionitrile

3-Methoxypropionitrile ≥98.0% (GC)
Synonym(s):
3-Methoxypropionitrile
Linear Formula:
CH3OCH2CH2CN
CAS Number:
Molecular Weight:
85.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing