Skip to Content
Merck
(3-Chloropropyl)trimethoxysilane ≥97%

(3-Chloropropyl)trimethoxysilane

(3-Chloropropyl)trimethoxysilane ≥97%
Synonym(s):
3-(Chloropropyl)-trimethoxysilane
Linear Formula:
Cl(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
198.72

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing