3,5-Dihydroxybenzyl alcohol
Synonym(s):
5-(Hydroxymethyl)resorcinol
Linear Formula:
(HO)2C6H3CH2OH
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing