Skip to Content
Merck
(2S)-(+)-Glycidyl tosylate 98%

(2S)-(+)-Glycidyl tosylate

(2S)-(+)-Glycidyl tosylate 98%
Synonym(s):
(S)-(+)-Glycidyl p-toluenesulfonate, (S)-(+)-Oxirane-2-methanol p-toluenesulfonate
Empirical Formula (Hill Notation):
C10H12O4S
CAS Number:
Molecular Weight:
228.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing