Skip to Content
Merck
2-Octyl-4-isothiazolin-3-one PESTANAL®, analytical standard

2-Octyl-4-isothiazolin-3-one

2-Octyl-4-isothiazolin-3-one PESTANAL®, analytical standard
Synonym(s):
2-Octyl-3(2H)-isothiazolone, Octhilinone
Empirical Formula (Hill Notation):
C11H19NOS
CAS Number:
Molecular Weight:
213.34
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing