Skip to Content
Merck
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline ≥99%

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline ≥99%
Synonym(s):
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, EEDQ, Ethyl 1,2-dihydro-2-ethoxyquinoline-1-carboxylate
Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing