2,3-Epoxypropyl methacrylate
Synonym(s):
Glycidyl methacrylate, 2,3-Epoxypropyl methacrylate, Methacrylic acid 2,3-epoxypropyl ester
Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing