Skip to Content
Merck
21-Hydroxyprogesterone ≥97% (HPLC)

21-Hydroxyprogesterone

21-Hydroxyprogesterone ≥97% (HPLC)
Synonym(s):
11-Deoxycorticosterone, 11-Desoxycorticosterone, 21-Hydroxy-4-pregnene-3,20-dione, 4-Pregnen-21-ol-3,20-dione, Cortexone, DOC, Desoxycortone, Kendall’s desoxy compound B, ‘Reichstein Q’
Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing