(1S)-(+)-(10-Camphorsulfonyl)oxaziridine
Synonym(s):
(1S)-(+)-(Camphorylsulfonyl)oxaziridine, (1S)-(+)-2,N-Epoxy-exo-10,2-bornanesultam
Empirical Formula (Hill Notation):
C10H15NO3S
CAS Number:
Molecular Weight:
229.30
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing