1-Chlorohexane
Synonym(s):
Hexyl chloride, 1-Chlorohexane
Linear Formula:
CH3(CH2)5Cl
CAS Number:
Molecular Weight:
120.62
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing