![1-Butyl-2,3-dimethylimidazolium tetrafluoroborate for electrochemistry, ≥99.0% (HPLC/T)](/deepweb/assets/sigmaaldrich/product/structures/890/098/f3746b0a-d464-4938-a4a6-2d1ed58939d5/640/f3746b0a-d464-4938-a4a6-2d1ed58939d5.png)
1-Butyl-2,3-dimethylimidazolium tetrafluoroborate
![1-Butyl-2,3-dimethylimidazolium tetrafluoroborate for electrochemistry, ≥99.0% (HPLC/T)](/deepweb/assets/sigmaaldrich/product/structures/890/098/f3746b0a-d464-4938-a4a6-2d1ed58939d5/640/f3746b0a-d464-4938-a4a6-2d1ed58939d5.png)
Empirical Formula (Hill Notation):
C9H17BF4N2
CAS Number:
Molecular Weight:
240.05
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing