Skip to Content
Merck
17α-Methyltestosterone VETRANAL®, analytical standard

17α-Methyltestosterone

17α-Methyltestosterone VETRANAL®, analytical standard
Synonym(s):
17α-Methyl-4-androsten-17β-ol-3-one, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing