Skip to Content
Merck
1,6-Anhydro-β-D-glucose analytical standard

1,6-Anhydro-β-D-glucose

1,6-Anhydro-β-D-glucose analytical standard
Synonym(s):
Levoglucosan
Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing