Skip to Content
Merck
1,2-Dioleoyl-sn-glycerol ≥97%

1,2-Dioleoyl-sn-glycerol

1,2-Dioleoyl-sn-glycerol ≥97%
Synonym(s):
(9Z)-9-Octadecenoic acid 1,1′-[(1S)-1-(hydroxymethyl)-1,2-ethanediyl] ester, (S)-1,2-Diolein, (S)-Glycerol 1,2-dioleate, 1,2-Di(cis-9-octadecenoyl)-sn-glycerol, DG(18:1(9Z)/18:1(9Z)/0:0), DG(18:1/18:1/0:0)
Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing