Skip to Content
Merck
1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride 97%

1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride

1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride 97%
Synonym(s):
Trimetazidine dihydrochloride
Empirical Formula (Hill Notation):
C14H22N2O3 · 2HCl
CAS Number:
Molecular Weight:
339.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing