Skip to Content
Merck
1-(1-Adamantyl)ethylamine hydrochloride 99%

1-(1-Adamantyl)ethylamine hydrochloride

1-(1-Adamantyl)ethylamine hydrochloride 99%
Synonym(s):
α-Methyltricyclo[3.3.1.13,7]-decane-1-methanamine hydrochloride, Rimantadine hydrochloride
Empirical Formula (Hill Notation):
C12H21N · HCl
CAS Number:
Molecular Weight:
215.76

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing