![1-[1-(2-Methoxyethyl)piperidin-3-yl]methanamine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/243/057/bcf8b7c9-b138-4c35-a26d-e45e6f4840cb/640/bcf8b7c9-b138-4c35-a26d-e45e6f4840cb.png)
1-[1-(2-Methoxyethyl)piperidin-3-yl]methanamine
![1-[1-(2-Methoxyethyl)piperidin-3-yl]methanamine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/243/057/bcf8b7c9-b138-4c35-a26d-e45e6f4840cb/640/bcf8b7c9-b138-4c35-a26d-e45e6f4840cb.png)
Empirical Formula (Hill Notation):
C9H20N2O
CAS Number:
Molecular Weight:
172.27
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing