Product Name
(2,3-Difluorophenyl)thiourea 97%
Product Number
654833
Product Brand
aldrich
CAS Number
572889-25-9
Molecular Weight
188.20
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE |
POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 43.1% - 46.2% |
NITROGEN 14.3% - 15.3% | |
SULFUR 16.4% - 17.7% | |
HIGH PRESSURE LIQUID | 96.5% (MINIMUM) |
APPROVED MARCH 9, 2005 BTR |