SY3H94E1CB1C
Synthonix Corporation
3,5-Difluorophenylzinc bromide, 0.50 M in THF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
bromo(3,5-difluorophenyl)zinc
Inchi Code
1S/C6H3F2.BrH.Zn/c7-5-2-1-3-6(8)4-5;;/h2-4H;1H;/q;;+1/p-1
InChI key
YZASGIRQZDSRKZ-UHFFFAOYSA-M
storage temp.
Refrigerator
Purity
0.50
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Solution
Product Link
https://synthonix.com/73186.html
Signal Word
Danger
Hazard Statements
MSDS
https://synthonix.com/73186.html
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service