Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11501

Copper naphthenate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 1 g

manufacturer/tradename

Chem Service, Inc. PS-2028

technique(s)

AAS: suitable
ICP: suitable

format

neat

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service