Skip to Content
Merck
All Photos(1)

Key Documents

442430

Supelco

1-Methylnaphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH3
CAS Number:
Molecular Weight:
142.20
Beilstein:
506793
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

autoignition temp.

984 °F

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.615 (lit.)

bp

240-243 °C (lit.)

mp

−22 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

Cc1cccc2ccccc12

InChI

1S/C11H10/c1-9-5-4-7-10-6-2-3-8-11(9)10/h2-8H,1H3

InChI key

QPUYECUOLPXSFR-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Kulka et al.
Mutation research, 208(3-4), 155-158 (1988-07-01)
Chromosome analyses were carried out in human lymphocytes treated in vitro with 1- and 2-methylnaphthalene (1-MN, 2-MN) in the presence and absence of the mammalian metabolic activation system, S9 mix. Without S9 mix there was no indication of induction of
Mykola Seredych et al.
Langmuir : the ACS journal of surfaces and colloids, 26(1), 227-233 (2009-12-30)
Two synthetic, polymer-derived carbons were modified with urea to incorporate nitrogen surface functional groups. Then they were investigated as adsorbents of dibenzothiophene (DBT) and 4, 6-dimethyldibenzothiophene (DMDBT) from simulated diesel fuel under dynamic conditions with the total concentration of sulfur
R E Rasmussen et al.
Journal of applied toxicology : JAT, 6(1), 13-20 (1986-02-01)
Male Swiss-Webster mice were exposed to naphthalene, 1-methyl-, 2-methyl-, 1-nitro- and 2-nitronaphthalene by intraperitoneal injection of peanut oil solutions over a dose range of 0.5-3.0 mmol kg-1 body weight. Treated mice were killed at times from 6 hours to 14
K G Kropp et al.
Biodegradation, 7(3), 203-221 (1996-06-01)
Dimethylbenzothiophenes are among the sulfur heterocycles in petroleum that are known to be degraded by microbial activity. Six of the 15 possible isomers of dimethylbenzothiophene were synthesized and used in bio-transformation studies with three Pseudomonas isolates that oxidize a variety
C E Cerniglia et al.
Applied and environmental microbiology, 47(1), 111-118 (1984-01-01)
Cunninghamella elegans metabolized 1- and 2-methylnaphthalene primarily at the methyl group to form 1- and 2-hydroxymethylnaphthalene, respectively. Other compounds isolated and identified were 1- and 2-naphthoic acids, 5-hydroxy-1-naphthoic acid, 5-hydroxy-2-naphthoic acid, 6-hydroxy-2-naphthoic acid, and phenolic derivatives of 1- and 2-methylnaphthalene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service