Skip to Content
Merck
All Photos(1)

Documents

SML0144

Sigma-Aldrich

2002-H20

≥98% (HPLC)

Synonym(s):

4-amino-N-[3-(2-benzoxazolyl)-4-hydroxyphenyl]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15N3O3
CAS Number:
Molecular Weight:
345.35
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)C(=O)Nc2ccc(O)c(c2)-c3nc4ccccc4o3

InChI

1S/C20H15N3O3/c21-13-7-5-12(6-8-13)19(25)22-14-9-10-17(24)15(11-14)20-23-16-3-1-2-4-18(16)26-20/h1-11,24H,21H2,(H,22,25)

InChI key

NVVIQGMTHCUMFT-UHFFFAOYSA-N

Biochem/physiol Actions

2002-H20 is an Alheimer′s Aβ peptide binder. It inhibits Aβ-induced death of PC12 cells, reducing the amount of toxic oligomer by enhancing fibril formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service