Skip to Content
Merck
All Photos(1)

Key Documents

SAB2103430

Sigma-Aldrich

Anti-SOCS7, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NAP4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 473.00

CHF 473.00


Estimated to ship on22 April 2025



Select a Size

Change View
100 μL
CHF 473.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 473.00


Estimated to ship on22 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

mouse, bovine, rat, human, pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOCS7(30837)

Immunogen

Synthetic peptide directed towards the N terminal region of human SOCS7

Biochem/physiol Actions

SOCS7 regulates signaling cascades probably through protein ubiquitination and/or sequestration. SOCS7 functions in insulin signaling and glucose homeostasis through IRS1 ubiquitination and subsequent proteasomal degradation. SOCS7 inhibits also prolactin, growth hormone and leptin signaling by preventing STAT3 and STAT5 activation, sequestering them in the cytoplasm and reducing their binding to DNA. SOCS7 may be a substrate recognition component of a SCF-like E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.

Sequence

Synthetic peptide located within the following region: LDPKALPPGLALERTWGPAAGLEAQLAALGLGQPAGPGVKTVGGGCCPCP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beibei Fu et al.
Cell death & disease, 11(9), 803-803 (2020-09-27)
Acute liver failure (ALF) is a rare but life-threatening systemic disorder. The innate immune regulation has an important role in this process; however, the specific mechanisms are not completely clear. Using the LPS + D-GalN-induced ALF mouse model, we found that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service