Skip to Content
Merck
All Photos(3)

Key Documents

SAB1404598

Sigma-Aldrich

Monoclonal Anti-CDC14A antibody produced in mouse

clone 1F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

cdc14, hCDC14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μG
CHF 474.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Estimated to ship on27 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC14A(8556)

General description

The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms. (provided by RefSeq)

Immunogen

CDC14A (NP_003663, 431 a.a. ~ 530 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PFRLSSSLQGSAVTLKTSKMALSPSATAKRINRTSLSSGATVRSFSINSRLASSLGNLNAATDDPENKKTSSSSKAGFTASPFTNLLNGSSQPTTRNYPE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service