Skip to Content
Merck
All Photos(1)

Documents

N6909

Sigma-Aldrich

Neurturin human

>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

NRTN, NTN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

15-50 ng/mL ED50

mol wt

monomer calculated mol wt ~12.5 kDa
predicted mol wt ~14 kDa by SDS-PAGE (reducing)

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NRTN(4902)

Biochem/physiol Actions

Member of the glial cell line-derived neurotrophic factor family of ligands. Exerts potent effects on the function and survival of central and peripheral neurons. Activates the RET receptor tyrosine kinase, MAP kinase, and phosphatidylinositol 3-kinase pathways. Human and mouse NTN share 91% sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA containing 1.25 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to bind recombinant human GFRα2/Fc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service