Skip to Content
Merck
All Photos(1)

Key Documents

M9557

Sigma-Aldrich

Anti-Macrophage Inflammatory Protein-3β antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-MIP-3β

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 437.00

CHF 437.00


Estimated to ship on28 April 2025



Select a Size

Change View
100 μG
CHF 437.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 437.00


Estimated to ship on28 April 2025


biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunocytochemistry: 5-15 μg/mL
neutralization: 0.8-4 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ccl19(24047)

General description

The antibody neutralizes the biological activity of recombinant mouse MIP-3β. Using 50 times more, the antibody will neutralize the biological activity of recombinant human MIP-3β. The antibody shows 5% cross-reactivity with recombinant human MIP-3β.

Immunogen

Recombinant mouse MIP-3β, expressed in Escherichia coli.

Biochem/physiol Actions

Macrophage Inflammatory Protein-3β is a chemokine that facilitates the migration of dendritic cells and antigen-presenting cells. It interacts with specific G protein-coupled seven-transmembrane domain chemokine receptor, CCR7 and promotes interactions between T cells and dendritic cells and migration of activated B cells to lymphoid tissues.

Physical form

Lyophilized from a 0.2 μm-filtered solution in phosphate-buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-mei Hou et al.
Acta pharmacologica Sinica, 30(3), 355-363 (2009-03-06)
Tumor immunotherapy aims at activating the body's own immune system to fight an existing tumor. Effective antitumor responses require tumor antigens to be presented to lymphocytes. We aimed to test the hypothesis that intratumoral administration of recombinant adenovirus encoding MIP3beta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service