Skip to Content
Merck
All Photos(1)

Key Documents

L3794

Sigma-Aldrich

LOK (1-348), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PRO2729, STK10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2-4 nmol/min·mg

mol wt

~65 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK10(6793)

General description

LOK (lymphocyte-oriented kinase) belongs to the Ste20 (sterile 20) family of serine/threonine kinases. It is expressed in lymphocytes, brain, colon, thymus, kidney, liver, small intestine and lung. LOK is commonly referred to as STK10 (serine/threonine protein kinase 10).

Biochem/physiol Actions

Mutations in LOK (lymphocyte-oriented kinase) are linked with the anti-apoptotic effects leading to carcinogenesis. It functions as an ERM (ezrin-radixin-moesin) kinase and thereby regulates cytoskeletal organization and lymphocyte migration. It can phosphorylate PLK1 (polo-like kinase 1).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazutaka Fukumura et al.
Oncology reports, 30(4), 1542-1548 (2013-07-12)
Peripheral T-cell lymphoma (PTCL) is an aggressive lymphoma with a 5-year overall survival rate of <30%. To identify carcinogenesis-related genes in PTCL, we conducted high-throughput resequencing of target-captured cDNA in a PTCL specimen, revealing a total of 19 missense mutations
Sarah A Walter et al.
The Journal of biological chemistry, 278(20), 18221-18228 (2003-03-18)
The Ste20 family of serine/threonine kinases plays an important role in numerous cellular functions such as growth, apoptosis, and morphogenesis. We have identified a previously cloned but uncharacterized family member termed Stk10, which is a human homolog of murine Lok
Natalya V Belkina et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(12), 4707-4712 (2009-03-04)
ERM (ezrin-radixin-moesin) proteins mediate linkage of actin cytoskeleton to plasma membrane in many cells. ERM activity is regulated in part by phosphorylation at a C-terminal threonine, but the identity of ERM kinases is unknown in lymphocytes and incompletely defined in
Li Tao et al.
The Biochemical journal, 363(Pt 1), 175-182 (2002-03-21)
T-cell activation requires signals from both the T-cell receptor (TcR) and other co-stimulatory molecules such as CD28. TcR- and CD28-mediated signals are integrated during T-cell activation resulting in the expression of cytokine genes such as interleukin-2 (IL-2). An enhancer element

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service