Skip to Content
Merck
All Photos(1)

Key Documents

I9149

Sigma-Aldrich

Inhibin from porcine ovary

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

Porcine

Quality Level

form

lyophilized powder

packaging

pkg of 2,000 units/vial
pkg of 1 vial

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibins are glycoproteins made of an α subunit and a β subunit. They suppress FSH (follicle stimulating hormone) secretion from the pituitary and also participate in control of gonadal functions.

Quantity

2,000 I.U. per vial

Physical form

Lyophilized from a buffered solution containing sodium chloride, bovine serum albumin, and trehalose.

Analysis Note

The biological activity is assayed according to the method of Tsonis et al., 1986 (J. Endocrinol. 110, 341) using WHO, 86/690 inhibin reference material as a standard.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Romagnani et al.
Kidney international, 83(3), 351-353 (2013-03-01)
Cultures of stem or progenitor cells have made critical contributions to the comprehension of tissue regeneration. In the kidney, primary cultures of human tubular progenitors became available only recently and allow dissection of the functional properties of tubular progenitors vs.
Sifrance Tran et al.
Journal of pediatric surgery, 48(1), e51-e54 (2013-01-22)
A 10 year old female presented with a 3 week history of persistent vaginal bleeding, lower abdominal pain, and generalized fatigue. Further work-up determined that she had severe autoimmune hypothyroidism, precocious puberty, and a large left cystic ovarian mass, consistent
N Iu Raĭgorodskaia et al.
Urologiia (Moscow, Russia : 1999), (2)(2), 88-92 (2013-06-26)
The article presents the results of a clinical evaluation of 39 boys with hypospadias aged 10-15 years. The examination included clinical assessment of pubertal development, and Doppler ultrasound scan of the gonads, transrectal ultrasound of the prostate in adolescents over
C Brömel et al.
Journal of veterinary internal medicine, 27(1), 76-82 (2013-01-15)
Studies in humans identified the synthesis and secretion of inhibin from adrenocortical tumors, but not pheochromocytoma (PHEO). Inhibin has not been examined in dogs as a serum biomarker for adrenal gland tumors. To determine serum inhibin concentration in dogs with
Brandon W Alleman et al.
American journal of obstetrics and gynecology, 208(6), 472-472 (2013-03-19)
The objective of the study was to create a predictive model for preterm birth (PTB) from available clinical data and serum analytes. Serum analytes and routine pregnancy screening plus cholesterol and corresponding health information were linked to birth certificate data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service