Skip to Content
Merck
All Photos(5)

Key Documents

HPA014836

Sigma-Aldrich

Anti-MBOAT2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Membrane-bound O-acyltransferase domain-containing protein 2, Anti-O-acyltransferase domain-containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

TQRRKNTHENIQLSQSKKFDEGENSLGQNSFSTTNNVCNQNQEIASRHSSLKQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBOAT2(129642)

General description

MBOAT2 (membrane bound O-acyltransferase domain containing 2) is a member of the LPCAT (lyso-PC acyltransferase) family of proteins, and is called LPCAT4. It is an acyltransferase, which is membrane-bound. In mice, MBOAT2 mRNA is predominantly expressed in brain, epididymis, testis, and ovary. Its C-terminal contains the ER (endoplasmic reticulum)-localization motif KKXX.

Immunogen

Membrane-bound O-acyltransferase domain-containing protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MBOAT2 (membrane bound O-acyltransferase domain containing 2) is responsible for the phosphatidylcholine (PC), and prefers fatty acid (FA)(16:1) and FA(18:1) as substrates. It is up-regulated in colorectal cancer, and this is linked with increase in PC. It also has (LPEAT) (lyso-phosphoethanolamine acyltransferase) activity and shows preference for 18:1-CoA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73261

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuya Kurabe et al.
Cancer science, 104(10), 1295-1302 (2013-07-03)
The identification of cancer biomarkers is critical for target-linked cancer therapy. The overall level of phosphatidylcholine (PC) is elevated in colorectal cancer (CRC). To investigate which species of PC is overexpressed in colorectal cancer, an imaging mass spectrometry was performed
Hideo Shindou et al.
Journal of lipid research, 50 Suppl, S46-S51 (2008-10-22)
Cells of all organisms are enclosed by a plasma membrane containing bipolar lipids, cholesterol, and proteins. Cellular membranes contain several classes of glycerophospholipids, which have numerous structural and functional roles in cells. Polyunsaturated fatty acids including arachidonic acid and eicosapentaenoic
Hideo Shindou et al.
The Journal of biological chemistry, 284(1), 1-5 (2008-08-23)
Cell membranes contain several classes of glycerophospholipids, which have numerous structural and functional roles in the cells. Polyunsaturated fatty acids, including arachidonic acid and eicosapentaenoic acid, are located at the sn-2 (but not sn-1)-position of glycerophospholipids in an asymmetrical manner.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service