Documents
EP5820707003
Eppendorf® Wide-mouth Bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.82
material
polypropylene bottle
polypropylene cap
square bottle
feature
graduations
with cap
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5820707003
capacity
500 mL
General description
500 mL wide-neck bottle which are plastic vessels for preparing, transporting, storing and centrifuging solid and liquid samples and reagents. Can be centrifuged in aresol-tight conditions and is autoclavable (121C, 20 mins). The centrifugation stability of consumables generally depends on the following conditions:
  • Properties of the consumable (e.g., material, shape)
  • Combination of centrifuge, rotor and, if applicable, adapter
  • Accuracy of fit of the consumable in the rotor bore or adapter
  • Centrifugation parameters (speed/g-force, temperature, centrifugation time)
  • Overall weight of consumable and contents
  • Physical and chemical properties of the centrifuged liquid
Legal Information
Eppendorf is a registered trademark of Eppendorf AG