Skip to Content
Merck
All Photos(1)

Documents

SK-N-F1

94092304, human nerve (bone marrow metastasis), Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SK-N-F1, 94092304

biological source

human nerve (bone marrow metastasis)

growth mode

Adherent/aggregates in suspension

karyotype

Aneuploid

morphology

Epithelial-like

products

Not specified

receptors

Transferrin receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human neuroblastoma

Cell Line Description

SK-N-F1 has been obtained from a bone marrow metastasis of a male patient with neuroblastoma. The poorly differentiated and slow growing cells secrete different neuroblastoma specific peptides and synthesise choline acetyltransferase. Reactivity of anti-leukaemia-associated and anti-foetal brain-associated monoclonal antibodies could be shown. Epithelial-like and aggregate-forming populations.

Application

Carcinogenesis studies, cytotoxicity assays.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 11,14
D16S539: 11,12
D5S818: 12,13
D7S820: 8,9
THO1: 6,9.3
TPOX: 8,9
vWA: 16,17

Culture Medium

DMEM + 4mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 2-4x10,000 cells/ml using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Clumped cells have tendency to float.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service