Skip to Content
Merck
All Photos(1)

Key Documents

I-10

89031605, mouse testis, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

I-10, 89031605

biological source

mouse testis

growth mode

Adherent

karyotype

2n = 40, hyperdiploid

morphology

Epithelial

products

Steroids

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c Leydig cell testicular tumour

Cell Line Description

Derived from a Leydig cell testicular tumour of a BALB/c mouse. The cells secrete steroids; this increases at saturation density. Secretion should be retained for over 50 passages. Exposure to 0.00001M cAMP stimulates secretion 4-5 fold. The major products are progesterone and its derivative 20 α-hydroxy-4-pregnen-3-one. It does not respond to interstitial cell stimulating hormone (ICSH) but is stimulated by cyclic adenosine monophosphate (AMP).

Application

Tumourigenicity studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 6x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service