Skip to Content
Merck
All Photos(1)

Key Documents

B2432

Sigma-Aldrich

8-Bromoadenosine-3′,5′-cyclic monophosphorothioate, Rp-isomer

≥98% (HPLC), solid

Synonym(s):

Rp-8-Br-cAMPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrN5O5PSNa
Molecular Weight:
446.15
UNSPSC Code:
41106305
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

packaging

vial of 5 μmol

solubility

water: soluble

storage temp.

−20°C

Biochem/physiol Actions

Competitive inhibitor of cyclic AMP-dependent protein kinase A (PKA) type I and II. Membrane-permeable analog of the cAMP inhibitor Rp-cAMPS (Prod. No. A-165).

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B T Gjertsen et al.
The Journal of biological chemistry, 270(35), 20599-20607 (1995-09-01)
Novel (Rp)-cAMPS analogs differed widely in ability to antagonize cAMP activation of pure cAMP-dependent protein kinase I and II and to antagonize actions of cAMP on gene expression, shape change, apoptosis, DNA replication, and protein phosphorylation in intact cells. These
Angeline Eymery et al.
Development (Cambridge, England), 143(15), 2767-2779 (2016-06-19)
Oocytes develop the competence for meiosis and early embryogenesis during their growth. Setdb1 is a histone H3 lysine 9 (H3K9) methyltransferase required for post-implantation development and has been implicated in the transcriptional silencing of genes and endogenous retroviral elements (ERVs).
Ho Lin et al.
Endocrinology, 150(1), 396-403 (2008-08-30)
The roles of cyclin-dependent kinase 5 (Cdk5) in central nervous system and neurodegenerative diseases have been intensely investigated in recent decades. Because protein expressions of Cdk5 and its regulator, p35, have been identified in Leydig cells, it is informative to
Mei-Chih Chen et al.
American journal of physiology. Cell physiology, 299(2), C516-C527 (2010-05-14)
The signaling mechanisms underlying cell differentiation have been extensively studied with the use of rat PC12 cells as a model system. Nerve growth factor (NGF) is a trophic factor inducing PC12 cell differentiation through the activation of the p35/cyclin-dependent kinase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service