Skip to Content
Merck
All Photos(2)

Key Documents

AV34590

Sigma-Aldrich

Anti-METTL3 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Methyltransferase like 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 354.00

CHF 354.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
CHF 354.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 354.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

dog, human, bovine, goat, mouse, horse, rat, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... METTL3(56339)

General description

Methyltransferase like 3 (METTL3 or M6A) codes for a 70 kDa subunit of MT-A. This protein forms a heterodimeric complex with METTL14 and modulates the methylation of the mammalian nuclear RNA, N6-adenosine.
Rabbit Anti-METTL3 antibody recognizes bovine, human, mouse, rat, zebrafish, and canine METTL3.

Immunogen

Synthetic peptide directed towards the middle region of human METTL3

Application

Rabbit Anti-METTL3 antibody is suitable for western blot applications at a concentration of 5 μg/ml.

Biochem/physiol Actions

METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.

Sequence

Synthetic peptide located within the following region: AKEPAKKSRKHAASDVDLEIESLLNQQSTKEQQSKKVSQEILELLNTTTA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianzhao Liu et al.
Nature chemical biology, 10(2), 93-95 (2013-12-10)
N(6)-methyladenosine (m(6)A) is the most prevalent and reversible internal modification in mammalian messenger and noncoding RNAs. We report here that human methyltransferase-like 14 (METTL14) catalyzes m(6)A RNA methylation. Together with METTL3, the only previously known m(6)A methyltransferase, these two proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service