Skip to Content
Merck
All Photos(1)

Documents

00033

Sigma-Aldrich

D-Arabinonic acid sodium salt

≥98.0% (TLC)

Synonym(s):

D-Arabonic acid sodium salt, Sodium D-arabinonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NaO6
CAS Number:
Molecular Weight:
188.11
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(O[Na])=O

InChI

1S/C5H10O6.Na/c6-1-2(7)3(8)4(9)5(10)11;/h2-4,6-9H,1H2,(H,10,11);/q;+1/p-1/t2-,3-,4+;/m1./s1

InChI key

IDBPBTCQINMQJK-PSRPMNHMSA-M

Biochem/physiol Actions

Metabolite of the D-arabinose pathway

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Ping Gai et al.
Plant, cell & environment, 37(6), 1474-1490 (2013-12-18)
To analyse the molecular mechanisms of phytoplasma pathogenicity, the comprehensive metabolomic changes of mulberry leaf and phloem sap in response to phytoplasma infection were examined using gas chromatography-mass spectrometry. The metabolic profiles obtained revealed that the metabolite compositions of leaf
B Lu et al.
Electrophoresis, 17(2), 325-332 (1996-02-01)
A rapid and sensitive method for the analysis of sugars was developed by capillary zone electrophoresis (CZE) with indirect UV detection. A mixture of nine mono- and oligosaccharides was separated in about 20 minutes on a fused silica capillary in
Osao Adachi et al.
Bioscience, biotechnology, and biochemistry, 75(9), 1801-1806 (2011-09-08)
In our previous study, a new microbial reaction yielding 4-keto-D-arabonate from 2,5-diketo-D-gluconate was identified with Gluconacetobacter liquefaciens RCTMR 10. It appeared that decarboxylation and dehydrogenation took place together in the reaction. To analyze the nature of the reaction, investigations were
Oxidation of D-glucose with oxygen in alkaline methanol-water mixtures: a convenient method of producing crystalline sodium D-arabinonate
Vuorinen, T., et al.
Starch, 43, 194-198 (1991)
George Cooper et al.
Journal of chromatography. A, 1216(40), 6838-6843 (2009-09-01)
Analysis of compounds in meteorites revealed a need to simultaneously characterize multiple enantiomers of sugar acids (aldonic acids) present in trace amounts. Analyses by gas chromatography-mass spectrometry demonstrated that all but two of the three-carbon through six-carbon straight-chained sugar acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service