Skip to Content
Merck
All Photos(1)

Key Documents

78039

Millipore

Bacillus ID Membrane

suitable for microbiology, pale pink colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
CHF 64.20

CHF 64.20


Estimated to ship on24 April 2025


Request more information

Select a Size

Change View
25 DISCS
CHF 64.20

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

CHF 64.20


Estimated to ship on24 April 2025


Request more information

membrane size

70 mm

Quality Level

sterility

sterile

form

disc

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Bacillus spp.

General description

Bacillus ID Membranes are substrate-carrying ID membranes used for rapid identification and differentiation of Bacillus sp. through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on a specific enzyme-substrate reaction in which Bacillus sp. will form green-colored colonies.

Application

Bacillus ID Membrane is used for rapid detection, confirmation, and differentiation of various species of Bacillus such as B. subtilis, B. cereus, B. thuringiensis, B. megaterium, B. coagulans, B. pumilus in food, meat, fish samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Bacillus cereus (10876) - Light blueBacillus subtilis (6633) - Yellowish green to greenBacillus thuringiensis (10876) - Light blue

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service