Skip to Content
Merck
All Photos(1)

Key Documents

77536

Supelco

Poncirin

analytical standard

Synonym(s):

(S)-5,7-Dihydroxy-4′-methoxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], 4′-O-Methylnaringin, Isosakuranetin-7-neohesperidoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34O14
CAS Number:
Molecular Weight:
594.56
Beilstein:
74301
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

SMILES string

COc1ccc(cc1)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)cc3O2

InChI

1S/C28H34O14/c1-11-21(32)23(34)25(36)27(38-11)42-26-24(35)22(33)19(10-29)41-28(26)39-14-7-15(30)20-16(31)9-17(40-18(20)8-14)12-3-5-13(37-2)6-4-12/h3-8,11,17,19,21-30,32-36H,9-10H2,1-2H3/t11-,17-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1

InChI key

NLAWPKPYBMEWIR-SKYQDXIQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poncirin is a flavanone glycoside, which is isolated from the water extract of fruits of Poncirus trifoliata. It is known to exhibit pharmacological properties such as anti-bacterial 
and anti-inflammatory activity.

Application

Poncirin (with other flavonoids of the bitter orange) was tested as a control agent for mosquito infestation, and was found to be larvacidal, to repel females from egg-laying on treated water, and to repel insects from biting topically treated human volunteers.
Poncirin may be used as a reference standard for the determination of poncirin in orange fruit extract using high-performance liquid chromatography coupled with diode array detector (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anti-Helicobacter pylori activity of the metabolites of poncirin from Poncirus trifoliata by human intestinal bacteria.
Kim B-J, et al.
Biological & Pharmaceutical Bulletin, 22(4), 422-424 (1999)
Inhibition of LPS-induced iNOS, COX-2 and cytokines expression by poncirin through the NF-kappaB inactivation in RAW 264.7 macrophage cells.
Kim B-J, et al.
Biological & Pharmaceutical Bulletin, 30(12), 2345-2351 (2007)
Poncirin promotes osteoblast differentiation but inhibits adipocyte differentiation in mesenchymal stem cells.
Yoon Y-H, et al.
European Journal of Pharmacology, 664(1-3), 54-59 (2011)
Determination of flavanones in orange juices obtained from different sources by HPLC/DAD.
Silva CRCL, et al.
Journal of Analytical Methods in Chemistry, 2014(1-3), 54-59 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service